Learn More
SOHLH1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524402
Catalog No. PIPA524402
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Probable transcription factor required during spermatogenesis and oogenesis.
Specifications
Specifications
SOHLH1
Polyclonal
Unconjugated
SOHLH1
Liquid
SOHLH1
KLH conjugated synthetic peptide between 40-68 amino acids from the N-terminal region of human SOHLH1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q5JUK2
C9orf157, NOHLH, TEB2, bA100C15.3, bHLHe80, newborn ovary helix loop helix, spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
402381