Learn More
SOHLH2 Mouse anti-Human, Clone: 1E1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522081
Catalog No. PIMA522081
Add to cart
Description
Description
Peptide Sequence: MASSIICQEH CQISGQAKID ILLVGDVTVG YLADTVQKLF ANIAEVTITI SDTKEAAALL DDCIFNMVLL KVPSSLSAEE LEAIKLIRFG KKKNTHSLFV FIIPENFKGC ISGHGMDIAL TEPLTMEKMS NVVKYWTTCP SNTVKTENAT GPEELGLPLQ RSYSEHLGYF PTDLFACSES LRNGNGLELN ASLSEFEKNK KISLLHSSKE KLRRLYRKHS SFCFW
Probable transcription factor, which may be involved in spermatogenesis and oogenesis.
Specifications
Specifications
SOHLH2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9NX45
TEB1, bHLHe81, spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 2
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
54937
ELISA, Immunoprecipitation, Western Blot
1E1
Unconjugated
SOHLH2
Liquid
SOHLH2
SOHLH2 (AAH25383, 1 a.a. ∽ 225 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human