Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC17A3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA124430

Catalog No. PIPA124430

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

SLC17A3 is a member of solute carrier family.

PA1-24430 detects Solute Carrier Family 17, Member 2 in Human samples.PA1-24430 has been successfully used in Western Blot procedures.PA1-24430 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human SLC17A3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Solute Carrier Family 17, Member 2
Polyclonal
Unconjugated
O00476
Rabbit
Antigen affinity chromatography
RUO
10786
Store at -20°C, Avoid Freeze/Thaw Cycles
Purified
Immunohistochemistry, Western Blot
1mg/mL
SLC17A3
SLC17A3
Synthetic peptide region of human SLC17A3.
50 μg
Primary
Human
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit