Learn More
Somatostatin Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA516253
Catalog No. PIPA516253
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-16253 targets Somatostatin in IHC (P) applications and shows reactivity with Human samples. The PA5-16253 immunogen is synthetic cyclic (1-14) somatostatin.
The cyclic polypeptide hormone Somatostatin was originally isolated from the hypothalamus and characterized by its ability to inhibit release of growth hormone from the pituitary gland. It exists in two forms, somatostatin-14, composed of 14 amino acids and somatostatin-28, a prohormone composed of 28 residues.
Specifications
Specifications
Somatostatin
Polyclonal
whole serum diluted in PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P61278
SST
Synthetic cyclic (1-14) somatostatin
500μL
4° C
Primary
6750
Immunohistochemistry (Paraffin)
Unconjugated
SST
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Human