Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHH Rat anti-Mouse, Clone: 171018, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523883

Catalog No. PIMA523883


Add to cart

Description

Description

In Western blots, approximately 50% cross-reactivity with recombit mouse (rm) Dhh(N-terminus; aa 23-198) is observed and no cross-reactivity with rmIhh (N-terminus; aa 66-240), rmDhh (C-terminus; aa 199-396), or rmShh (C-terminus; aa 199-437) is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

Sonic Hedgehog, which is expressed only during embryogenesis, is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the Sonic Hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved, the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the developing embryo. Defects in this protein or in its signalling pathway are a cause of holoprosencephaly (HPE), a disorder in which the developing forebrain fails to correctly separate into right and left hemispheres. HPE is manifested by facial deformities. In addition, it is thought that mutations in this gene or in its signalling pathway may be responsible for VACTERL syndrome, which is characterized by vertebral defects, anal atresia, tracheoesophageal fistula with esophageal atresia, radial and renal dysplasia, cardiac anomalies, and limb abnormalities.
Specifications

Specifications

SHH
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q62226
TPT, HHG1, HLP3, HPE3, SMMCI, TPTPS, MCOPCB5, SHH
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
20423
ELISA, Immunohistochemistry, Neutralization, Western Blot
171018
Unconjugated
Shh
Lyophilized
SHH
E. coli-derived recombit mouse Sonic Hedgehog Cys25-Gly198 (Lys122Arg)
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit