Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorcin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528359

Catalog No. PIPA528359


Add to cart

Description

Description

PA5-28359 targets Sorcin in WB applications and shows reactivity with Human and mouse samples. The PA5-28359 immunogen is recombit fragment corresponding to a region within amino acids 1 and 198 of Human Sorcin. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Calcium-binding protein that modulates excitation-contraction coupling in the heart. Contributes to calcium homeostasis in the heart sarcoplasmic reticulum. Modulates the activity of RYR2 calcium channels.
Specifications

Specifications

Sorcin
Polyclonal
Unconjugated
SRI
Liquid
SRI
Recombit fragment corresponding to a region within amino acids 1 and 198 of Human Sorcin
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunoprecipitation, Western Blot
0.65 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P30626, Q6P069
CP22, SCN, 22 kDa protein, CP-22, H_RG167B05.1, V19, calcium binding protein amplified in mutlidrug-resistant cells
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
109552, 6717
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit