Learn More
sortilin-related receptor, L(DLR class) A repeats-containing, Mouse, Clone: 3F2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SORL1.
Supplier: Abnova Corporation H00006653M01
Description
This gene encodes a protein that belongs to the families of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins, of low density lipoprotein receptor (LDLR) proteins, and of fibronectin type III repeats proteins. In addition to VPS10, LDLR and fibronectin type 3 domains, this protein also includes an epidermal growth factor precursor-like module, a single transmembrane segment and a cytoplasmic tail with features similar to endocytosis- and sorting-competent receptors. Members of the VPS10 domain-containing receptor family are large with many exons but the CDS lengths are usually less than 3700 nt; this gene is an exception to the pattern with a CDS length greater than 6600 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. The encoded protein is mainly intracellular and localizes in the paranuclear compartment. It is synthesized as a preproprotein, and when the propeptide is still attached, no binding occurs to the VPS10 domain. This gene is strongly expressed in the central nervous system. [provided by RefSeq
Sequence: SAALQPEPIKVYGQVSLNDSHNQMVVHWAGEKSNVIVALARDSLALARPKSSDVYVSYDYGKSFKKISDKLNFGLGNRSEAVIAQFYHSPADNKRYIFADSpecifications
sortilin-related receptor, L(DLR class) A repeats-containing | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003105 | |
SORL1 | |
SORL1 (NP_003096, 82 a.a. ∼ 181 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
3F2 | |
Mouse monoclonal antibody raised against a partial recombinant SORL1. | |
SORL1 | |
C11orf32/FLJ21930/FLJ39258/LR11/LRP9/SORLA/SorLA-1/gp250 | |
Mouse | |
Affinity Purified | |
RUO | |
6653 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.