Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SOX1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145865

Catalog No. NBP145865

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SOX1
0.5 mg/mL
Western Blot
P53783
SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1
Rabbit
IgG
0.1 mg
Stem Cells
Primary
6656
Human, Mouse, Rat
Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
SOX1
Synthetic peptide corresponding to residues surrounding amino acids 322 of mouse Sox-1.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
The antibody recognizes ~ 43 kDa Sox-1 in samples of human and mouse origins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only