Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SOX17, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 120-170 of human SOX17 was used as the immunogen.
Antigen SOX17
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224644SS
View Documents
Novus Biologicals
NBP224644SS
0.025 mg
Each for $119.60
Add to cart
 
NBP224644
View Documents
Novus Biologicals
NBP224644
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

SOX17 Polyclonal antibody specifically detects SOX17 in Human, Mouse, Porcine, Chicken, Opossum, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
A portion of amino acids 120-170 of human SOX17 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q9H6I2
PBS, 0.05% BSA with 0.05% Sodium Azide
IgG
SOX17
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human,88%homologous inmonkey, and 62% homologous in mouse.
64321
0.5 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit