Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX18 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124474

Catalog No. PA124474


Add to cart

Description

Description

PA1-24474 detects SOX18 in Human samples. PA1-24474 has been successfully used in Western Blot procedures. PA1-24474 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human SOX18. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

SOX18 a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. This protein plays a role in hair, blood vessel, and lymphatic vessel development. Mutations in this gene have been associated with recessive and domit forms of hypotrichosis-lymphedema-telangiectasia.
Specifications

Specifications

SOX18
Polyclonal
Unconjugated
SOX18
Liquid
SOX18
Synthetic peptide region of human SOX18.
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P35713
HLTS
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
54345
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit