Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of SOX2.

Manufacturer:  Abnova Corporation PAB12735

Catalog No. 89-024-833


Add to cart

Description

Description

This intronless gene encodes a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of this gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. Mutations in this gene have been associated with optic nerve hypoplasia and with syndromic microphthalmia, a severe form of structural eye malformation. This gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). [provided by RefSeq

Specifications

Specifications

SOX2
Unconjugated
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
SOX2
ANOP3|MCOPS3|MGC2413
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes ~35 KDa of human SOX2.
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of SOX2.
In TBS, pH 7.2 (BSA, 10% Proclin300)
Liquid
SOX2
A synthetic peptide corresponding to N-terminus of human SOX2.
RUO
Primary
6657
Bovine, Canine, Chicken, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit