Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
$204.00 - $494.50
Specifications
Antigen | SOX2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5 ug/mL, Flow Cytometry 1:100, Immunohistochemistry 1:125 - 1:250, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:125 - 1:250 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11037235T
|
Novus Biologicals
NB11037235SS |
0.025 mL |
Each for $204.00
|
|
|||||
NB11037235
|
Novus Biologicals
NB11037235 |
0.1 mL |
Each for $494.50
|
|
|||||
Description
SOX2 Polyclonal specifically detects SOX2 in Human, Mouse, Rat, Canine, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SOX2 | |
Western Blot 0.5 ug/mL, Flow Cytometry 1:100, Immunohistochemistry 1:125 - 1:250, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:125 - 1:250 | |
Polyclonal | |
Rabbit | |
Cellular Markers, Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
PBS with 0.02% Sodium Azide | |
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
SOX2 | |
IgG | |
Affinity Purified | |
Human, sheep and mouse. 92% sequence identity with chicken, 86% sequence identity with Xenopus, and 71% sequence identity with Zebrafish proteins. |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Canine, Sheep | |
P48431 | |
6657 | |
A synthetic peptide made to the N-terminal region of human SOX2 protein (within residues 1-100). [Swiss-Prot# P48431] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title