Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SOX4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP150776

Catalog No. NBP150776


Add to cart

Description

Description

SOX4 Polyclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SOX4
0.25 mg/mL
Western Blot 2.5 - 5 ug/mL, ELISA
Q06945
ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4
Rabbit
IgG
0.1 mg
Stem Cell Markers, Stem Cells
Primary
6659
ELISA, Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA, 50% Glycerol with 0.03% Proclin
Affinity Purified
SOX4
Synthetic peptide selected from the N-terminal region of human SOX4
Affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only