Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox7 Mouse anti-Human, Unlabeled, Clone: S5-1216, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564694

Catalog No. BDB564694


Add to cart

Description

Description

The S5-1216 monoclonal antibody specifically binds to human Sox7, a member of the SRY-related HMG-box (SOX) family of transcription factors. Sox7 plays a key role in cardiac, hematopoietic and vascular development. Sox7 is also involved in endoderm lineage determination, where Sox7 is expressed in the extraembryonic endoderm but not in the definitive endoderm. Over-expression of Sox7 in human embryonic stem cells leads to differentiation into lineage restricted extraembryonic endoderm progenitors. Furthermore, Sox7 has a tumor suppressive activity, as indicated by its down-regulation in colon, prostate, lung and breast cancer.

Specifications

Specifications

Sox7
S5-1216
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
0.1mg
Store undiluted at 4°C
Monoclonal
Flow Cytometry
0.5mg/mL
Transcription factor SOX-7, SOX7
Affinity Purified
Human Sox7 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit