Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128072

Catalog No. PA128072


Add to cart

Description

Description

PA1-28072 detects SOX8 from human and mouse samples. PA1-28072 has been successfully used in immunohistochemistry, immunoprecipitation, and Western blot applications. The PA1-28072 immunogen is synthetic peptide: GDGHHHGDHTGQTHG conjugated to BSA, corresponding to amino acids 209-223 of Human SOX8. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. This protein may be involved in brain development and function. Haploinsufficiency for this protein may contribute to the mental retardation found in haemoglobin H-related mental retardation (ART-16 syndrome).
Specifications

Specifications

SOX8
Polyclonal
Unconjugated
SOX8
Liquid
SOX8
synthetic peptide GDGHHHGDHTGQTHG conjugated to BSA, corresponding to amino acids 209-223 of Human SOX8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.2
P57073, Q04886
MGC24837
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20681, 30812
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit