Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA19103
Description
This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology.
SP1 Polyclonal antibody specifically detects SP1 in Human samples. It is validated for Western Blot.Specifications
SP1 | |
Polyclonal | |
Unconjugated | |
SP1 | |
1110003E12Rik; AA450830; AI845540; HGNC:11205; SP1; Sp1 transcription factor; Sp1-1; specificity protein 1; trans-acting transcription factor 1; transcription factor Sp1; TSFP1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6667 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P08047 | |
SP1 | |
Synthetic peptide corresponding to residues CSRIESPNENSNNSQ of human SP1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction