Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPA17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA130328

Catalog No. PIPA130328

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-30328 detects SP17 from human samples. PA1-30328 has been successfully used in Western blot applications. The PA1-30328 immunogen is: Synthetic peptide (C terminal).

This gene encodes a protein present at the cell surface. The N-terminus has sequence similarity to human cAMP-dependent protein kinase A (PKA) type II alpha regulatory subunit (RIIa) while the C-terminus has an IQ calmodulin-binding motif. The central portion of the protein has carbohydrate binding motifs and likely functions in cell-cell adhesion. The protein was initially characterized by its involvement in the binding of sperm to the zona pellucida of the oocyte. Recent studies indicate that it is also involved in additional cell-cell adhesion functions such as immune cell migration and metastasis. A retrotransposed pseudogene is present on chromosome 10q22.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPA17
Polyclonal
Unconjugated
Q15506
SPA17
Synthetic peptide (C terminal).
1 mL
Primary
Human
Antibody
IgG
Western Blot
0.2 mg/mL
SPA17
CT22, SP17, SP17-1
Rabbit
Antigen affinity chromatography
RUO
53340
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit