Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526934

Catalog No. PIPA526934


Add to cart

Description

Description

SP5 binds to GC boxes promoters elements. SP5 probable transcriptional activator that has a role in the coordination of changes in transcription required to generate pattern in the developing embryo.
Specifications

Specifications

SP5
Polyclonal
Unconjugated
SP5
Liquid
SP5
KLH conjugated synthetic peptide between 369-398 amino acids from the C-terminal region of human SP5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q6BEB4, Q9JHX2
transcription factor Sp5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
389058, 64406
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit