Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPA-1 Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611214

 View more versions of this product

Catalog No. BDB611214


Add to cart

Description

Description

Trafficking of immune cells is essential for the development of an effective immune response. Immunocompetent lymphocytes recirculate through lymphoid tissues “in search of” their corresponding specific antigen. Antigen encounter triggers signaling cascades that result in gene activation and clonal expansion. Central to this process is the activation of Ras which is mediated by the conversion of GTP to GDP and involves multiple Ras GTPase-activating proteins (GAPs). Rap1, a member of the Ras family of GTPases, functions to inhibit the Ras-MAP kinase pathway. A GAP specific for Rap1, rap1GAP, has been identified in human brain. However, this GAP exhibits little expression in lymphoid tissues where elevated Rap1 is detected. SPA-1 exhibits a specific GAP activity for Rap1 and Rap2. It is highly expressed in lymphocytes and minimally expressed in tissues such as brain, kidney, and pancreas where abundant rap1GAP is detected. Human and mouse SPA-1 are 90% identical and contain proline-rich regions and a rap1GAP-related domain followed by a coiled-coil region. Thus, SPA-1 is a Rap specific GAP that functions with rap1GAP to regulate Rap1 and Ras mediated signaling.

Host Species: Mouse
Clone: 3
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human SPA-1 aa. 911-1027
Formula Weight [Chemical]: 130kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

SPA-1
3
Unconjugated
Murine
IgG1
RUO
Primary
Human, Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human SPA-1 aa. 911-1027
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.