Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARCL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523715

Catalog No. PIPA523715

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This is a secreted protein. Found in the extracellular matrix. It’s highly expressed in lymph node, brain, heart, lung, skeletal muscle, ovary, small intestine, and colon, with lower levels in placenta, pancreas ,testis, spleen, and thymus, and no expression in kidney, liver, and peripheral blood leukocytes.
Specifications

Specifications

SPARCL1
Polyclonal
PBS with 0.09% sodium azide
Q14515
PIG33, SC1, MAST 9, SPARC-like 1 (mast9, hevin), SPARC-like protein 1, hevin, high endothelial venule protein, mast9, proliferation-inducing protein 33
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8404
Immunohistochemistry, Western Blot
Unconjugated
SPARCL1
Liquid
SPARCL1
KLH conjugated synthetic peptide between 208-236 amino acids from the central region of human SPARCL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit