Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPCS3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513630

Catalog No. PIPA513630

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

SPCS3 is a component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
Specifications

Specifications

SPCS3
Polyclonal
PBS with 0.09% sodium azide
P61009, Q6ZWQ7
Signal peptidase complex subunit 3; Microsomal signal peptidase 23 kDa subunit; SPase 22 kDa subunit; SPC22/23; SPC22; SPCS3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60559, 76687
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SPCS3
Liquid
SPCS3
KLH conjugated synthetic peptide between 152-180 amino acids from the C-terminal region of human SPCS3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit