Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPDYE3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523861
Description
SPDYE3 Polyclonal antibody specifically detects SPDYE3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SPDYE3 | |
Polyclonal | |
Unconjugated | |
SPDYE3 | |
putative Speedy protein E3; Putative WBSCR19-like protein 7; SPDYE3; speedy; speedy homolog E3; speedy protein E3; speedy/RINGO cell cycle regulator family member E3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
441272 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
A6NKU9 | |
SPDYE3 | |
KLH conjugated synthetic peptide between 455-483 amino acids from the central region of human SPDYE3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction