Learn More
Spectrin beta-3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA146008
Catalog No. PA146008
Add to cart
Description
Description
This antibody is not recommended for Western blot analysis. Suggested positive control: HeLa whole cell lysate.
Mutations in spectrins are a previously unknown cause of ataxia and neurodegenerative disease that affect membrane proteins involved in glutamate signaling. Spectrin-beta IIIs have been recognized as ataxia disease genes and their mutations cause spinocerebellar ataxia type 5 (SCA5).
Specifications
Specifications
Spectrin beta-3
Polyclonal
Unconjugated
SPTBN2
Liquid
SPTBN2
Residues 2150 and 2200 of human spectrin, beta, non-erythrocytic 2 (spinocerebellar ataxia 5 ).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
tris citrate, tris phosphate with 0.09% sodium azide; pH 7-8
O15020
GTRAP41, SCA5
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
6712