Learn More
Sp56 Mouse anti-Hamster, Mouse, Clone: 12D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110870
Catalog No. 01-671-879
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10870 detects mouse Sperm Protein sp56 from mouse and hamster samples. MA1-10870 has been successfully used in ELISA, Western blot, and immunohistochemistry applications. The MA1-10870 immunogen is a mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
Specifications
Specifications
Sp56
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Q60736
AC122773.1, AV264399, SP56
Mouse
IgG3
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
22789
ELISA, Immunohistochemistry, Western Blot
12D2
Unconjugated
Zp3r
Liquid
ZP3R
Mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
Protein G
RUO
Antibody
Monoclonal
Hamster, Mouse