Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAG16 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525198

Catalog No. PIPA525198


Add to cart

Description

Description

Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively.
Specifications

Specifications

SPAG16
Polyclonal
PBS with 0.09% sodium azide
Q8N0X2, Q8K450
PF20, WDR29, WD repeat domain 29, pf20 protein homolog, sperm-associated WD repeat protein, sperm-associated antigen 16 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
66722, 79582
Immunohistochemistry
Unconjugated
SPAG16
Liquid
SPAG16
KLH conjugated synthetic peptide between 276-304 amino acids from the central region of human SPG16
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit