Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112764
Description
SAMD8 Polyclonal antibody specifically detects SAMD8 in Human samples. It is validated for ELISA, Western Blot.Specifications
SAMD8 | |
Polyclonal | |
Unconjugated | |
SAMD8 | |
1110053F04Rik; 1700010P07Rik; Ceramide phosphoethanolamine synthase; CPE synthase; epididymis luminal protein 177; epididymis secretory sperm binding protein Li 181mP; FLJ25082; GrailEXP prediction; HEL-177; HEL-S-181mP; RP11-487I5.5; SAM domain-containing protein 8; SAMD8; SMSr; sphingomyelin synthase related; sphingomyelin synthase-related protein 1; sterile alpha motif domain containing 8; sterile alpha motif domain-containing 8; sterile alpha motif domain-containing protein 8 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
142891 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
Q96LT4 | |
SAMD8 | |
Full-length recombinant sphingomyelin synthase r fusion protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction