Learn More
SPHK1 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA112856
Catalog No. 01-675-837
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A suggested positive control for this product is rat kidney lysate.
Sphingosine Kinase 1 (Sphk1) catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
Specifications
Specifications
SPHK1
Polyclonal
PBS with 0.08% sodium azide
Q91V26
SPHK1
Synthetic peptide derived from rat sphingosine kinase 1, long form
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Western Blot
Unconjugated
Sphk1
Liquid
Rabbit
IgG
10 Miniblots
-80° C, Avoid Freeze/Thaw Cycles
Primary
170897