Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPHK1 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112885

Catalog No. 01-675-855


Add to cart

Description

Description

Sphingosine Kinase 1 (Sphk1) catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
Specifications

Specifications

SPHK1
Polyclonal
PBS with 0.08% sodium azide
Q91V26
SPHK1
Synthetic peptide derived from rat sphingosine kinase 1, long form
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Rat
Western Blot
Unconjugated
Sphk1
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
170897
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit