Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPHK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522994

Catalog No. PIPA522994


Add to cart

Description

Description

Sphingosine kinase 1 (SPHK1) catalyzes the phosphorylation of sphingosine to yield sphingosine-1-phosphate (SPP). SPP functions as a lipid signaling molecule that mediates both intra- and extra-cellular responses. SPHK1 activity may play a role in several cellular processes such as apoptisis, proliferation, inflammation, and migration.
Specifications

Specifications

SPHK1
Polyclonal
Unconjugated
SPHK1
Liquid
SPHK1
The epitope recognized by this antibody maps to a region between residue 334-384 of human sphingosine kinase 1 using the numbering given in entry NP_001136073.1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunoprecipitation, Western Blot
1 mg/mL
tris citrate with 0.09% sodium azide; pH 7-8
Q9NYA1
SPHK, SK 1, SPK 1
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
8877
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit