Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Sphingosine Kinase 1/SPHK1, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP122974AF700

Catalog No. NBP122974Z


Add to cart

Description

Description

Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Sphingosine Kinase 1/SPHK1
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
SPHK1
sphingosine kinase 1
42.5 kDa
0.1 ml
Lipid and Metabolism
Primary
8877
Western Blot
Western Blot
Q2M3C7
EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only