Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Rabbit anti-Human, Rat, DyLight 405, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP122974V
Description
Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human, Rat samples. It is validated for Western Blot.Specifications
Sphingosine Kinase 1/SPHK1 | |
1mg/mL | |
Western Blot | |
Q2M3C7 | |
EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
RUO | |
Store at 4C in the dark. | |
Polyclonal | |
Human, Rat |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Affinity Purified | |
SPHK1 | |
sphingosine kinase 1 | |
42.5 kDa | |
0.1 ml | |
Lipid and Metabolism | |
Primary | |
8877 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only