Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPHK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528099
Description
Recommended positive controls: A431, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SPHK1 Polyclonal antibody specifically detects SPHK1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SPHK1 | |
Polyclonal | |
Unconjugated | |
SPHK1 | |
1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8877 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.17 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9NYA1 | |
SPHK1 | |
Synthetic peptide corresponding to a region within amino acids 321 and 384 of Human SPHK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction