Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinesin Rat anti-Mouse, Clone: 282324, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523983

Catalog No. PIMA523983


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit human Spinesin is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes.
Specifications

Specifications

Spinesin
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9ER04
SPINESIN, transmembrane protease serine 5
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
80893
Immunoprecipitation, Western Blot
282324
Unconjugated
Tmprss5
Lyophilized
TMPRSS5
Mouse myeloma cell line NS0-derived recombit mouse Spinesin Tyr61-Arg445
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit