Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPIRE2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524099

Catalog No. PIPA524099

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in vesicle transport processes providing a novel link between actin organization and intracellular transport.
Specifications

Specifications

SPIRE2
Polyclonal
PBS with 0.09% sodium azide
Q8WWL2, Q8K1S6
Spir-2, protein spire homolog 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
234857, 84501
Western Blot
Unconjugated
SPIRE2
Liquid
SPIRE2
KLH conjugated synthetic peptide between 285-313 amino acids from the central region of human SPIRE2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit