Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human SPOCK1 protein.

Manufacturer:  Abnova Corporation H00006695B02

Catalog No. 89-006-274


Add to cart

Description

Description

This gene encodes the protein core of a seminal plasma proteoglycan containing chondroitin- and heparan-sulfate chains. The protein's function is unknown, although similarity to thyropin-type cysteine protease-inhibitors suggests its function may be related to protease inhibition. [provided by RefSeq

Sequence: MPAIAVLAAAAAAWCFLQVESRHLDALAGGAGPNHGNFLDNDQWLSTVSQYDRDKYWNRFRDDDYFRNWNPNKPFDKALDPSKDPCLKVKCSPHKVCVTQDYQTALCVSRKHLLPRQKKGNVAQKHWVGPSNLVKCKPCPVAQSAMVCGSDGHSYTSKCKLEFHACSTGKSLATLCDGPCPCLPEPEPPKHKAERSACTDKELRNLASRLKDWFGALHEDANRVIKPTSSNTAQGRFDTSILPICKDSLGWMFNKLDMNYDLLLDPSEINAIYLDKYEPCIKPLFNSCDSFKDGKLSNNEWCYCFQKPGGLPCQNEMNRIQKLSKGKSLLGAFIPRCNEEGYYKATQCHGSTGQCWCVDKYGNELAGSRKQGAVSCEEEQETSGDFGSGGSVVLLDDLEYERELGPKDKEGKLRVHTRAVTEDDEDEDDDKEDEVGYIW
Specifications

Specifications

sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1
Unconjugated
No additive
BC030691.2
FLJ37170|SPOCK|TESTICAN|TIC1
Mouse
50 uL
Cell Adhesion
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human SPOCK1 protein.
SPOCK1
Serum
SPOCK1
SPOCK1 (AAH30691.1, 1 a.a. ~ 439 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6695
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit