Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPON1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA129480

Catalog No. PIPA129480

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-29480 detects SPON1 from rat, human, mouse samples.PA1-29480 has been successfully used in Western blot applications.The PA1-29480 immunogen is: Synthetic peptide, corresponding to amino acids 549-807 of SPON1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPON1
Polyclonal
SPON1
KIAA0762, MGC10724, f-spondin
Chicken
Antigen affinity chromatography
RUO
10418
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
Q9HCB6, Q8VCC9, P35446
SPON1
Synthetic peptide, corresponding to amino acids 549-807 of SPON1.
50 μg
Primary
Human, Mouse, Rat
Antibody
IgY
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit