Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPOP, Polyclonal, Invitrogen™

Catalog No. PIPA127695
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127695 50 μg
1 options

Catalog No. PIPA127695

Supplier: Thermo Scientific PA127695

Chicken Polyclonal Antibody

Inhibits IPF1/PDX1 transactivation of established target promoters. such as insulin. may be by recruiting a repressor complex (By similarity). In complex with CUL3. involved in ubiquitination of both PCGF4 and H2AFY.This protein is a protein that may modu

PA1-27695 detects SPOP from human, mouse, and rat samples.PA1-27695 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Speckle-type POZ protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPOP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene SPOP
Gene Accession No. O43791, Q6ZWS8
Gene Alias TEF2
Gene Symbols SPOP
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Speckle-type POZ protein.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8405
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.