Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524688

Catalog No. PIPA524688


Add to cart

Description

Description

SPRN has PrP(C)-like neuroprotective activity. SPRN may act as a modulator for the biological actions of normal and abnormal PrP.
Specifications

Specifications

SPRN
Polyclonal
PBS with 0.09% sodium azide
Q5BIV9, Q8BWU1
SHADOO, SHO, bA108K14.1, hypothetical protein BC004409, protein shadoo, shadow of prion protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
212518, 503542
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPRN
Liquid
SPRN
KLH conjugated synthetic peptide between 76-105 amino acids from the C-terminal region of human SPRN
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit