Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRR1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526568
Description
SPRR1A Polyclonal antibody specifically detects SPRR1A in Human samples. It is validated for Western Blot.Specifications
SPRR1A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P35321 | |
SPRR1A | |
KLH conjugated synthetic peptide between 61-89 amino acids from the C-terminal region of human SPRR1A | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SPRR1A | |
19 kDa pancornulin; AI528815; cornifin A; cornifin-A; mSPRR1A; small proline rich protein 1A; small proline-rich protein 1A; small proline-rich protein 1A-like; small proline-rich protein 1B (cornifin); small proline-rich protein IA; Spr; SPR1 A; Spr1a; SPR-IA; SPRK; Sprr; Sprr1; Sprr1a; Sprr1al; Sprr1b | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
6698 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction