Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525418

Catalog No. PIPA525418

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of SPT13 remains unknown.
Specifications

Specifications

SPATA13
Polyclonal
PBS with 0.09% sodium azide
Q96N96
ARHGEF29, ASEF2, APC-stimulated guanine nucleotide exchange factor 2, adenomatous polyposis coli stimulated exchange factor 2, spermatogenesis-associated protein 13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
221178
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SPATA13
Liquid
SPATA13
KLH conjugated synthetic peptide between 204-232 amino acids from the central region of human SPT13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit