Learn More
SR-BI Rabbit anti-Hamster, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA131393
Catalog No. PA131393
Add to cart
Description
Description
PA1-31393 detects SR-BI from rat, hamster, and mouse samples. PA1-31393 has been successfully used in immunoprecipitation and Western blot applications. The PA1-31393 immunogen is synthetic peptide: C-SPAAKGTVLQEAKL conjugated to KLH, corresponding to amino acids 496-509 of Mouse SR-BI. Store at 4°C short term. For extended storage, aliquot and store at -20°C or below. Avoid freeze/thaw cycles.
Specifications
Specifications
SR-BI
Polyclonal
PBS with 0.02% sodium azide
Q8WTV0, Q61009, P97943
SCARB1, SR-B1, SR-BI, Scavenger Receptor-BI, Scavenger Receptor-B1
Rabbit
IgG
RUO
Antibody
Polyclonal
Hamster, Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
SCARB1
Liquid
SCARB1
synthetic peptide C-SPAAKGTVLQEAKL conjugated to KLH, corresponding to amino acids 496-509 of Mouse SR-BI.
100 μL
4° C, do not freeze
Primary
100736552, 20778, 25073, 949