Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRBD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524530

Catalog No. PIPA524530

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SRBD1
Polyclonal
PBS with 0.09% sodium azide
Q8N5C6
H_NH0576F01.1, S1 RNA-binding domain-containing protein 1, WUGSC:H_NH0576F01.1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55133
Immunohistochemistry, Western Blot
Unconjugated
SRBD1
Liquid
SRBD1
KLH conjugated synthetic peptide between 934-963 amino acids from the C-terminal region of human SRBD1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit