Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC-1 Mouse, Unlabeled, Clone: 8, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612378

Catalog No. BDB612378


Add to cart

Description

Description

Signal transduction via nuclear hormone receptors is important for cell growth and differentiation, development, and homeostasis. Nuclear hormone receptors are ligand-activated transcription factors that modulate target gene expression.These ligand/receptor complexes also interact with transcriptional coactivators which enhance ligand-dependent transcription. Various classes of coactivators have been identified, including SRC-1 and its related proteins, such as TIF-2/GRIP-1, RIP140, AIB-1, and TIF-1α and -1β. SRC-1 is a member of the basic helix-loop-helix-PAS domain family. It contains an N-terminal basic-helix-loop-helix (bHLH), two N-terminal PAS domains, and a glutamine-rich region near the C-terminus. SRC-1 interacts with many different steroid receptors and recruitment of SRC-1 to promoter sites may facilitate opening of chromatin structure via its histone acetyltransferase activity, as well as stabilization of the preinitiation complex. ERK-2 can phosphorylate SRC-1 at threonine and serine sites in vitro. In addition, SRC-1 expression in cells stimulated with EGF enhances
progesterone receptor-mediated activation of a target reporter gene. Thus, SRC-1 is an important transcriptional co-activator involved in steroid receptor signaling.

Immunofluorescence, Western Blotting

Specifications

Specifications

SRC-1
8
Unconjugated
Affinity Purified
Human SRC-1 aa. 761-863
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit