Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Src Mouse anti-Avian, Human, Mouse, Mink, Rat, Clone: GD11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05184
Description
Anti-Src Antibody, clone GD11 detects level of Src and has been published and validated for use in immunoprecipitation and western blotting.Specifications
Src | |
Monoclonal | |
Unconjugated | |
NM_005417.3; NM_198291.1 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunoprecipitation, Western Blot | |
GD11 | |
Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. | |
SRC, c-src, p60-Src, SRC1, c-SRC, ASV, pp60c-src | |
Recombinant chicken pp60src; v-src | |
200 μg | |
Avian, Human, Mink, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only