Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC Family Negative Regulatory (pY) Site Rabbit anti-Human, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen 44912

Catalog No. 44-912


Add to cart

Description

Description

This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the maligt progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene. Transcript Variant: This variant (1) represents the longer transcript. Both isoforms 1 and 2 encode the same protein.
Specifications

Specifications

SRC Family Negative Regulatory (pY) Site
Polyclonal
Dulbecco′s PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
P12931
EC 2.7.10.2, p60-Src, c-Src, pp60c-src
Rabbit
IgG
100 μL
-20°C
Primary
6714
Western Blot
Unconjugated
SRC
Liquid
SRC
synthesized phosphopeptide derived from the region of human Hck that contains tyrosine 522.
Purified
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit