Learn More
Src Family (WTAPE) Mouse anti-Human, Mouse, Rat, Clone: 2E8.2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051461
Catalog No. 05-146-1MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-Src Family (WTAPE) Antibody, clone 2E8.2 (Mouse Monoclonal Antibody) validated in western blotting to detect Src Family (WTAPE) also known as Integrin-linked kinase, 59 kDa serine/threonine-protein kinase.
Specifications
Specifications
Src Family (WTAPE)
2E8.2
Protein G purified mouse monoclonal in PBS and 0.05% NaN3.
Purified
Murine
IgG3, κ, κ
200μg
Primary
Human, Murine, Rat
Western Blot
Unconjugated
NP_005408
pp60c-src, p60-Src, c-Src
Shared Src family sequence found at the C-term. Amino acid sequence GAKFPIKWTAPE.
Protein G purfied
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal