Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRD5A2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP136990
Description
SRD5A2 Polyclonal specifically detects SRD5A2 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
SRD5A2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
3-oxo-5-alpha-steroid 4-dehydrogenase 2,5 alpha-SR2, EC 1.3.99.5, MGC138457, S5AR 2, SR type 2, Steroid 5-alpha-reductase 2,3-oxo-5 alpha-steroid 4-dehydrogenase 2, steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 2), Type II 5-alpha reductase | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific to the C Terminus. Cross reacts with Rat and Human. Expected from sequence similarity: Dog, Pig. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_000339.2 | |
SRD5A2 | |
Peptide with sequence C-KMFEDYPKSRK corresponding to C-Terminus according to NP_000339.2. | |
0.1 mg | |
Cancer | |
6716 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction