Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146142

Catalog No. PA146142


Add to cart

Description

Description

This antibody is predicted to react with porcine based on 100% sequence homology. This antibody has 94% sequence homology with bovine, 88% sequence homology with zebrafish and 70% sequence homology with chicken. Suggested positive control: human liver lysate.

SREBP (Sterol Regulatory Element Binding Protein) 1 and 2 are transcription factors which participate in the control of cholesterol homeostasis. SREBP proteins, which are attached to the endoplasmic reticulum and nuclear envelope, are proteolytically cleaved and thus activated in response to conditions of low cellular sterol. SREBPs may also play some role in the apoptotic processes
Specifications

Specifications

SREBP1
Polyclonal
Unconjugated
SREBF1
Liquid
SREBF1
Human SREBP1 protein sequence (between residues 700-800) [UniprotNo. P36956].
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse, Rat
Western Blot
1.1 mg/mL
tris glycine with 150mM NaCl and 0.05% sodium azide
P36956, Q9WTN3, P56720
SREBP-1c, SREBP1, bHLHd1, nt-SREBP1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100689018, 20787, 6720, 78968
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit