Learn More
MilliporeSigma™ anti-SRp30b (SC35), Polyclonal,
Manufacturer:  MilliporeSigma™ 061364
Catalog No. 06-136-4MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
SRp30b (SC35)
Unconjugated
Purified
Rabbit
Protein A
2-8°C for 12 months
Polyclonal
Human
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
SC-35; PR264; SFRS2; SC35; SRp30b; SFRS2A; Protein PR2643; SRSF2
Linear peptide corresponding to human SRp30b.
100μL
Primary
NP_001182356