Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRP54 Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610941

 View more versions of this product

Catalog No. BDB610941


Add to cart

Description

Description

Ribosomes that exist freely in the cytosol or those attached to the ER are intrinsically the same in their translational properties. ER-bound ribosomes are responsible for the production of secretory proteins and integral ER, Golgi, lysosomal, and plasma membrane spanning proteins. Such proteins contain signal sequences that direct their synthesis to the ER membrane. As the nascent polypeptide emerges from the ribosome, a signal recognition particle (SRP) binds to the signal sequence and serves to couple the ribosome to the protein-translocating machinery in the ER membrane. Although the SRP is a 325kDa ribonucleoprotein, its 54kDa subunit (SRP54) mediates interaction with, and targeting of, the nascent protein to the ER. Via its C-terminal M-domain, SRP54 associates with the nascent protein and inhibits its elongation. This complex binds to the SRP receptor on the ER, the ribosome is delivered to the translocation machinery, SRP is released, and elongation resumes. Targeting and insertion are tightly coupled to a GTPase cycle that involves SRP54 and SRP receptor. Although the mechanisms are unclear, release of SRP from the ER-bound complex requires GTP hydrolysis.

Immunofluorescence, Western Blotting

Specifications

Specifications

SRP54
30
Unconjugated
Affinity Purified
Human SRP54 aa. 262-476
150μg
Cell Biology
Primary
Canine, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.